Learn More
Fisherbrand™ Raised Shelves for Stainless Steel Tanks
Product Code. 17571906 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Fisher 12 L Dig.Heated Circ. Bath
Fisher 18 L Dig.Heated Circ. Bath
Fisher 26 L Dig.Heated Circ. Bath
Fisher 38 L Dig.Heated Circ. Bath
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17571906 Fisher 12 L Dig.Heated Circ. Bath Each
17581906 Fisher 18 L Dig.Heated Circ. Bath Each
17591906 Fisher 26 L Dig.Heated Circ. Bath Each
17501916 Fisher 38 L Dig.Heated Circ. Bath Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17571906
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Raised shelves for use with stainless steel tanks.
  • For use with digital heated circulating baths
  • Reversible
  • Allows for two shelf depths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fisher 12 L Dig.Heated Circ. Bath
Type Raised Shelf