Learn More
Grant Instruments™ Raised Shelf for Unstirred Water Baths
Product Code. 11406487
Change view
Click to view available options
Type:
Shelf for 12 L Tanks
Shelf for 18 L Tanks
Shelf for 26 L Tanks
Shelf for 34 L Tanks
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11406487 Shelf for 18 L Tanks Each
11496477 Shelf for 12 L Tanks Each
11416487 Shelf for 26 L Tanks Each
11426487 Shelf for 34 L Tanks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11406487 Supplier Grant Instruments™ Supplier No. RS22
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Grant Heated Circulating Baths
Type Shelf for 18 L Tanks
Height (Metric) 40 or 135 mm