missing translation for 'onlineSavingsMsg'
Learn More

RAF709, MedChemExpress

Product Code. p-200098501
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495787

Brand: Medchem Express HY100510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RAF709 is a potent, selective, and efficacious RAF inhibitor with IC50s of 0.4 nM and 0.5 nM for BRAF and CRAF, respectively. Antitumor efficacy.

RAF709 is a potent, selective, and efficacious RAF inhibitor with IC50s of 0.4 nM and 0.5 nM for BRAF and CRAF, respectively. Antitumor efficacy.

Specifications

Chemical Name or Material RAF709
CAS 1628838-42-5
Color White
Purity Grade Notes Research
Molecular Formula C28H29F3N4O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (184.31 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CN=C(C)C(C2=CC(N3CCOCC3)=C(OC4CCOCC4)N=C2)=C1)C5=CC=CC(C(F)(F)F)=C5
Molecular Weight (g/mol) 542.55
Formula Weight 542.55
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.