Learn More
RAF709, MedChemExpress
Product Code. 16495787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16495787 5 mg 5mg
16489616 50 mg 50mg
16454356 100 mg 100mg
16452596 25 mg 25mg
16433058 10 mg 10mg
5 options
Product Code. 16495787
Brand: Medchem Express HY100510
MedChemExpress RAF709 is a potent, selective, and efficacious RAF inhibitor with IC50s of 0.4 nM and 0.5 nM for BRAF and CRAF, respectively. Antitumor efficacy.
RAF709 is a potent, selective, and efficacious RAF inhibitor with IC50s of 0.4 nM and 0.5 nM for BRAF and CRAF, respectively. Antitumor efficacy.
Specifications
Chemical Name or Material RAF709
CAS 1628838-42-5
Color White
Purity Grade Notes Research
Molecular Formula C28H29F3N4O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (184.31 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CN=C(C)C(C2=CC(N3CCOCC3)=C(OC4CCOCC4)N=C2)=C1)C5=CC=CC(C(F)(F)F)=C5
Molecular Weight (g/mol) 542.55
Formula Weight 542.55
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease