Learn More
Rad23 Antibody (1E4D6) - BSA Free, R&D Systems™
Product Code. 18609856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609856 0.025 mg 0.03mg
18221131 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18609856 Supplier Novus Biologicals Supplier No. NBP2525500.025mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Rad23 Monoclonal specifically detects Rad23 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen hHR23A
Applications Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1E4D6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias hHR23A, HR23A
Gene Symbols RAD23A
Host Species Mouse
Immunogen Purified recombinant fragment of human RAD23A (AA: 1-363) expressed in E. Coli.
Molecular Weight of Antigen 40 kDa
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5886
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Mehr anzeigen Weniger anzeigen