Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Shop All Thermo Scientific ProductsDescription
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Color | Silver |
| Description | 76 4 mL/5 mL Ampules |
| For Use With (Equipment) | All CryoMed controlled-rate freezers |
| Holds | 76 4 mL/5 mL Ampules |
| Type | Vial Rack (Small) |