Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Shop All Thermo Scientific ProductsDescription
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Color | Black |
| Description | 162 Canes, 5 to 6 Ampules/Cane |
| For Use With (Equipment) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 |
| Holds | 162 Canes, 5 to 6 Ampules/Cane |
| Type | Cane Freezing Rack |