Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Product Code. 10265034 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
10 Canisters (for cord blood, Model 4000610)
161 1.2 mL/2 mL Ampules
161 4 mL/5 mL Ampules
162 Canes, 5 to 6 Ampules/Cane
3 x 4/5 mL Cryo Racks (Large)
3 x 4/5 mL Cryo Racks (Small)
40 0.25/0.5 mL Straws
48 Skin Packets (2 level)
5 x 1.2/2 mL Cryo Racks (Large)
5 x 1.2/2 mL Cryo Racks (Small)
76 1.2 mL/2 mL Ampules
76 4 mL/5 mL Ampules
77 Canes
Adjustable, 10-position
Type:
Cane Freezing Rack
Canister Rack
Skin Rack
Straw Rack
Vial Rack (Large)
Vial Rack (Small)
Vial Rack Holder
Unit Size:
Each
Product Code. Holds Type unitSize
10265034 48 Skin Packets (2 level) Skin Rack Each
15425204 Adjustable, 10-position Canister Rack Each
11736629 40 0.25/0.5 mL Straws Straw Rack Each
11726499 162 Canes, 5 to 6 Ampules/Cane Cane Freezing Rack Each
12770348 10 Canisters (for cord blood, Model 4000610) Canister Rack Each
11796489 77 Canes Cane Freezing Rack Each
10633575 76 1.2 mL/2 mL Ampules Vial Rack (Small) Each
10040524 5 x 1.2/2 mL Cryo Racks (Small) Vial Rack Holder Each
10365954 161 1.2 mL/2 mL Ampules Vial Rack (Large) Each
10244604 5 x 1.2/2 mL Cryo Racks (Large) Vial Rack Holder Each
11794898 76 4 mL/5 mL Ampules Vial Rack (Small) Each
11704908 3 x 4/5 mL Cryo Racks (Small) Vial Rack Holder Each
11714908 161 4 mL/5 mL Ampules Vial Rack (Large) Each
11724908 3 x 4/5 mL Cryo Racks (Large) Vial Rack Holder Each
14 options
Product Code. 10265034
Brand: Thermo Scientific™ 4000310
Only null left
Add to basket
Add to basket
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Description 48 Skin Packets (2 level)
For Use With (Equipment) All CryoMed controlled-rate freezers
Holds 48 Skin Packets (2 level)
Type Skin Rack