Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Product Code. 10040524 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
10 Canisters (for cord blood, Model 4000610)
161 1.2 mL/2 mL Ampules
161 4 mL/5 mL Ampules
162 Canes, 5 to 6 Ampules/Cane
3 x 4/5 mL Cryo Racks (Large)
3 x 4/5 mL Cryo Racks (Small)
40 0.25/0.5 mL Straws
48 Skin Packets (2 level)
5 x 1.2/2 mL Cryo Racks (Large)
5 x 1.2/2 mL Cryo Racks (Small)
76 1.2 mL/2 mL Ampules
76 4 mL/5 mL Ampules
77 Canes
Adjustable, 10-position
Type:
Cane Freezing Rack
Canister Rack
Skin Rack
Straw Rack
Vial Rack (Large)
Vial Rack (Small)
Vial Rack Holder
Unit Size:
Each
Product Code. Holds Type unitSize
10040524 5 x 1.2/2 mL Cryo Racks (Small) Vial Rack Holder Each
15425204 Adjustable, 10-position Canister Rack Each
11736629 40 0.25/0.5 mL Straws Straw Rack Each
11726499 162 Canes, 5 to 6 Ampules/Cane Cane Freezing Rack Each
10265034 48 Skin Packets (2 level) Skin Rack Each
12770348 10 Canisters (for cord blood, Model 4000610) Canister Rack Each
11796489 77 Canes Cane Freezing Rack Each
10633575 76 1.2 mL/2 mL Ampules Vial Rack (Small) Each
10365954 161 1.2 mL/2 mL Ampules Vial Rack (Large) Each
10244604 5 x 1.2/2 mL Cryo Racks (Large) Vial Rack Holder Each
11794898 76 4 mL/5 mL Ampules Vial Rack (Small) Each
11704908 3 x 4/5 mL Cryo Racks (Small) Vial Rack Holder Each
11714908 161 4 mL/5 mL Ampules Vial Rack (Large) Each
11724908 3 x 4/5 mL Cryo Racks (Large) Vial Rack Holder Each
14 options
Product Code. 10040524
Brand: Thermo Scientific™ 4000702
Only null left
Add to basket
Add to basket
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Description Five 1.2 mL/2 mL Cryo Racks (Small)
For Use With (Equipment) All CryoMed controlled-rate freezers
Holds 5 x 1.2/2 mL Cryo Racks (Small)
Type Vial Rack Holder