Learn More
Thermo Scientific™ Specialty Racks
Product Code. 12938284 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Microplates
Microplates or Deepwell Microplates
Product Type:
Box Racks - Sliding Drawer
Side Access Freezer Rack (Adjustable)
Side Access Freezer Rack (Microplate/Deepwell)
Side Access Freezer Rack (Microplate/Locking Rod)
Specialty Rack - 8-Compartment Container
For Use With (Equipment):
13 cu. ft. Upright Freezers
17.3, 23 cu. ft. Upright Freezers
32 cu.ft. Revco Upright ULT
Thermo Scientific TDE and TSX Series (5 shelves)
Unit Size:
Each
Positions:
12 Boxes/Rack
15 Boxes/Rack
20 Boxes/Rack
22 Boxes/Rack
36 Plates/Rack
56 Plates/Rack
77 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
98 microplates with lids, 119 microplates without lids, and 35 deepwell
Product Code. Holds Product Type For Use With (Equipment) unitSize Positions
12938284 General Freezer Items Specialty Rack - 8-Compartment Container 13 cu. ft. Upright Freezers Each -
12662725 Microplates Side Access Freezer Rack (Microplate/Deepwell) 13 cu. ft. Upright Freezers Each 56 Plates/Rack
12672725 Microplates Side Access Freezer Rack (Microplate/Deepwell) 17.3, 23 cu. ft. Upright Freezers Each 84 Plates/Rack
10784964 Microplates Side Access Freezer Rack (Microplate/Deepwell) 17.3, 23 cu. ft. Upright Freezers Each 36 Plates/Rack
12054128 Boxes (2 in.) Side Access Freezer Rack (Adjustable) 13 cu. ft. Upright Freezers Each 15 Boxes/Rack
10133933 Boxes (3 in.) Side Access Freezer Rack (Adjustable) 17.3, 23 cu. ft. Upright Freezers Each 12 Boxes/Rack
10153693 Boxes (2 in.) Side Access Freezer Rack (Adjustable) 17.3, 23 cu. ft. Upright Freezers Each 20 Boxes/Rack
10022573 Boxes (2 in.) Side Access Freezer Rack (Adjustable) 17.3, 23 cu. ft. Upright Freezers Each 22 Boxes/Rack
11934257 Boxes (2 in.) Box Racks - Sliding Drawer 13 cu. ft. Upright Freezers Each -
12622725 Boxes (3 in.) Box Racks - Sliding Drawer 13 cu. ft. Upright Freezers Each 9 Boxes/Rack
10072043 Boxes (2 in.) Box Racks - Sliding Drawer 17.3, 23 cu. ft. Upright Freezers Each -
10082043 Boxes (3 in.) Box Racks - Sliding Drawer 17.3, 23 cu. ft. Upright Freezers Each 12 Boxes/Rack
11349321 Boxes (2 in.) Box Racks - Sliding Drawer Thermo Scientific TDE and TSX Series (5 shelves) Each 20 Boxes/Rack
11329291 Boxes (2 in.) Side Access Freezer Rack (Adjustable) Thermo Scientific TDE and TSX Series (5 shelves) Each 20 Boxes/Rack
11339291 Boxes (3 in.) Side Access Freezer Rack (Adjustable) Thermo Scientific TDE and TSX Series (5 shelves) Each 15 Boxes/Rack
15824314 Microplates or Deepwell Microplates Side Access Freezer Rack (Microplate/Deepwell) Thermo Scientific TDE and TSX Series (5 shelves) Each 77 Plates/Rack
11379311 Microplates or Deepwell Microplates Side Access Freezer Rack (Microplate/Locking Rod) 32 cu.ft. Revco Upright ULT Each 98 microplates with lids, 119 microplates without lids, and 35 deepwell
11339321 Boxes (3 in.) Box Racks - Sliding Drawer Thermo Scientific TDE and TSX Series (5 shelves) Each 15 Boxes/Rack
18 options
Product Code. 12938284
Brand: Thermo Scientific™ 830030
Only null left
Add to basket
Add to basket
Maximize the storage space in your ultra-low temperature freezer with these racks.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Holds General Freezer Items
Shape Square
For Use With (Equipment) 13 cu. ft. Upright Freezers
Width (English) 7 in.
Height (English) 5.3 in.
Depth (English) 18 in.
Depth (Metric) 45.7 cm
Product Type Specialty Rack - 8-Compartment Container
Shelves 3 Tiers
Height (Metric) 13.5 cm
Width (Metric) 18 cm