Learn More
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Product Code. 30280805 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
96-well Microplate Holder
Adjustable 15 mL Conical Tube Rack
Adjustable 50 mL Conical Tube Rack
Adjustable Tube Rack
Control Box
EU Plug
Fixed Conical Tube Rack
Fixed Tube Rack
General Power Adapter
Rubber Platform
Shaker Platform
Spring Platform
Spring Wire Rack
Sticky Mat
UK Plug
Universal Platform
Dimensions (L x W):
140 x 140 mm
158 x 95 mm
21 x 30 mm
24 x 30 mm
240 x 350 mm
27 x 22 mm
350 x 240 mm
360 x 300 mm
40 x 14 mm dia.
40 x 16 mm
40 x 16 mm dia.
40 x 18 mm
428 x 295 mm
45.7 x 45.7 cm
450 x 400 mm
720 x 440 mm
900 x 540 mm
N/A
Unit Size:
1 unit
Each
Product Code. Product Type Dimensions (L x W) unitSize
30280805 Adjustable 50 mL Conical Tube Rack 24 x 30 mm 1 unit
15391115 Fixed Tube Rack 40 x 14 mm dia. Each
17594865 Shaker Platform 45.7 x 45.7 cm Each
30280797 Fixed Tube Rack 27 x 22 mm 1 unit
15301125 Fixed Tube Rack 40 x 16 mm dia. Each
30280795 Adjustable Tube Rack 40 x 16 mm 1 unit
15311125 Adjustable Tube Rack 40 x 14 mm dia. Each
30280800 Adjustable Tube Rack 40 x 18 mm 1 unit
15321125 Adjustable Tube Rack 40 x 16 mm dia. Each
30280791 Adjustable Tube Rack 27 x 22 mm 1 unit
15331125 Universal Platform 360 x 300 mm Each
30280794 Adjustable 50 mL Conical Tube Rack 21 x 30 mm 1 unit
15341125 Rubber Platform N/A Each
15351125 96-well Microplate Holder N/A Each
30280798 Spring Platform 900 x 540 mm 1 unit
15361125 Spring Wire Rack 240 x 350 mm Each
30280790 Spring Platform 720 x 440 mm 1 unit
15371125 Sticky Mat 140 x 140 mm Each
30280793 Spring Platform 350 x 240 mm 1 unit
15381125 Control Box 158 x 95 mm Each
30280801 Spring Platform 450 x 400 mm 1 unit
15391125 General Power Adapter N/A Each
30280788 Spring Platform 428 x 295 mm 1 unit
30280789 Fixed Conical Tube Rack N/A 1 unit
30280799 Adjustable 50 mL Conical Tube Rack N/A 1 unit
15301135 EU Plug N/A Each
30280802 Adjustable 15 mL Conical Tube Rack N/A 1 unit
15311135 UK Plug N/A Each
28 options
Product Code. 30280805
Brand: Thermo Scientific™ 88881141
Only null left
Add to basket
Add to basket
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Adjustable 50 mL Conical Tube Rack
Dimensions (L x W) 24 x 30 mm
For Use With (Equipment) Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers