Learn More
- Home
- Products
- Shakers
- Shaker Accessories
- Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Product Code. 30280800 Shop All Thermo Scientific Products| Product Code. | Product Type | Dimensions (L x W) | unitSize |
|---|---|---|---|
| 30280800 | Adjustable Tube Rack | 40 x 18 mm | 1 unit |
| 15391115 | Fixed Tube Rack | 40 x 14 mm dia. | Each |
| 17594865 | Shaker Platform | 45.7 x 45.7 cm | Each |
| 30280797 | Fixed Tube Rack | 27 x 22 mm | 1 unit |
| 15301125 | Fixed Tube Rack | 40 x 16 mm dia. | Each |
| 30280795 | Adjustable Tube Rack | 40 x 16 mm | 1 unit |
| 15311125 | Adjustable Tube Rack | 40 x 14 mm dia. | Each |
| 15321125 | Adjustable Tube Rack | 40 x 16 mm dia. | Each |
| 30280791 | Adjustable Tube Rack | 27 x 22 mm | 1 unit |
| 15331125 | Universal Platform | 360 x 300 mm | Each |
| 30280794 | Adjustable 50 mL Conical Tube Rack | 21 x 30 mm | 1 unit |
| 15341125 | Rubber Platform | N/A | Each |
| 30280805 | Adjustable 50 mL Conical Tube Rack | 24 x 30 mm | 1 unit |
| 15351125 | 96-well Microplate Holder | N/A | Each |
| 30280798 | Spring Platform | 900 x 540 mm | 1 unit |
| 15361125 | Spring Wire Rack | 240 x 350 mm | Each |
| 30280790 | Spring Platform | 720 x 440 mm | 1 unit |
| 15371125 | Sticky Mat | 140 x 140 mm | Each |
| 30280793 | Spring Platform | 350 x 240 mm | 1 unit |
| 15381125 | Control Box | 158 x 95 mm | Each |
| 30280801 | Spring Platform | 450 x 400 mm | 1 unit |
| 15391125 | General Power Adapter | N/A | Each |
| 30280788 | Spring Platform | 428 x 295 mm | 1 unit |
| 30280789 | Fixed Conical Tube Rack | N/A | 1 unit |
| 30280799 | Adjustable 50 mL Conical Tube Rack | N/A | 1 unit |
| 15301135 | EU Plug | N/A | Each |
| 30280802 | Adjustable 15 mL Conical Tube Rack | N/A | 1 unit |
| 15311135 | UK Plug | N/A | Each |
Product Code. 30280800
Brand: Thermo Scientific™ 88881138C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
- Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
- Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
| Product Type | Adjustable Tube Rack |
| Dimensions (L x W) | 40 x 18 mm |
| For Use With (Equipment) | Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers |