Learn More
- Home
- Products
- Shakers
- Shaker Accessories
- Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Product Code. 30280794 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
96-well Microplate Holder
Adjustable 15 mL Conical Tube Rack
Adjustable 50 mL Conical Tube Rack
Adjustable Tube Rack
Control Box
EU Plug
Fixed Conical Tube Rack
Fixed Tube Rack
General Power Adapter
Rubber Platform
Shaker Platform
Spring Platform
Spring Wire Rack
Sticky Mat
UK Plug
Universal Platform
Dimensions (L x W):
140 x 140 mm
158 x 95 mm
21 x 30 mm
24 x 30 mm
240 x 350 mm
27 x 22 mm
350 x 240 mm
360 x 300 mm
40 x 14 mm dia.
40 x 16 mm
40 x 16 mm dia.
40 x 18 mm
428 x 295 mm
45.7 x 45.7 cm
450 x 400 mm
720 x 440 mm
900 x 540 mm
N/A
Unit Size:
1 unit
Each
| Product Code. | Product Type | Dimensions (L x W) | unitSize |
|---|---|---|---|
| 30280794 | Adjustable 50 mL Conical Tube Rack | 21 x 30 mm | 1 unit |
| 15391115 | Fixed Tube Rack | 40 x 14 mm dia. | Each |
| 17594865 | Shaker Platform | 45.7 x 45.7 cm | Each |
| 30280797 | Fixed Tube Rack | 27 x 22 mm | 1 unit |
| 15301125 | Fixed Tube Rack | 40 x 16 mm dia. | Each |
| 30280795 | Adjustable Tube Rack | 40 x 16 mm | 1 unit |
| 15311125 | Adjustable Tube Rack | 40 x 14 mm dia. | Each |
| 30280800 | Adjustable Tube Rack | 40 x 18 mm | 1 unit |
| 15321125 | Adjustable Tube Rack | 40 x 16 mm dia. | Each |
| 30280791 | Adjustable Tube Rack | 27 x 22 mm | 1 unit |
| 15331125 | Universal Platform | 360 x 300 mm | Each |
| 15341125 | Rubber Platform | N/A | Each |
| 30280805 | Adjustable 50 mL Conical Tube Rack | 24 x 30 mm | 1 unit |
| 15351125 | 96-well Microplate Holder | N/A | Each |
| 30280798 | Spring Platform | 900 x 540 mm | 1 unit |
| 15361125 | Spring Wire Rack | 240 x 350 mm | Each |
| 30280790 | Spring Platform | 720 x 440 mm | 1 unit |
| 15371125 | Sticky Mat | 140 x 140 mm | Each |
| 30280793 | Spring Platform | 350 x 240 mm | 1 unit |
| 15381125 | Control Box | 158 x 95 mm | Each |
| 30280801 | Spring Platform | 450 x 400 mm | 1 unit |
| 15391125 | General Power Adapter | N/A | Each |
| 30280788 | Spring Platform | 428 x 295 mm | 1 unit |
| 30280789 | Fixed Conical Tube Rack | N/A | 1 unit |
| 30280799 | Adjustable 50 mL Conical Tube Rack | N/A | 1 unit |
| 15301135 | EU Plug | N/A | Each |
| 30280802 | Adjustable 15 mL Conical Tube Rack | N/A | 1 unit |
| 15311135 | UK Plug | N/A | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
- Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
- Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
| Product Type | Adjustable 50 mL Conical Tube Rack |
| Dimensions (L x W) | 21 x 30 mm |
| For Use With (Equipment) | Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers |