Learn More
- Home
- Products
- Shakers
- Shaker Accessories
- Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Product Code. 15351125 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
96-well Microplate Holder
Adjustable 15 mL Conical Tube Rack
Adjustable 50 mL Conical Tube Rack
Adjustable Tube Rack
Control Box
EU Plug
Fixed Conical Tube Rack
Fixed Tube Rack
General Power Adapter
Rubber Platform
Shaker Platform
Spring Platform
Spring Wire Rack
Sticky Mat
UK Plug
Universal Platform
Dimensions (L x W):
140 x 140 mm
158 x 95 mm
21 x 30 mm
24 x 30 mm
240 x 350 mm
27 x 22 mm
350 x 240 mm
360 x 300 mm
40 x 14 mm dia.
40 x 16 mm
40 x 16 mm dia.
40 x 18 mm
428 x 295 mm
45.7 x 45.7 cm
450 x 400 mm
720 x 440 mm
900 x 540 mm
N/A
Unit Size:
1 unit
Each
28 product options available for selection
| Product Code. | Product Type | Dimensions (L x W) | Unit Size |
|---|---|---|---|
| 15351125 | 96-well Microplate Holder | N/A | Each |
| 15391115 | Fixed Tube Rack | 40 x 14 mm dia. | Each |
| 17594865 | Shaker Platform | 45.7 x 45.7 cm | Each |
| 30280797 | Fixed Tube Rack | 27 x 22 mm | 1 unit |
| 15301125 | Fixed Tube Rack | 40 x 16 mm dia. | Each |
| 30280795 | Adjustable Tube Rack | 40 x 16 mm | 1 unit |
| 15311125 | Adjustable Tube Rack | 40 x 14 mm dia. | Each |
| 30280800 | Adjustable Tube Rack | 40 x 18 mm | 1 unit |
| 15321125 | Adjustable Tube Rack | 40 x 16 mm dia. | Each |
| 30280791 | Adjustable Tube Rack | 27 x 22 mm | 1 unit |
| 15331125 | Universal Platform | 360 x 300 mm | Each |
| 30280794 | Adjustable 50 mL Conical Tube Rack | 21 x 30 mm | 1 unit |
| 15341125 | Rubber Platform | N/A | Each |
| 30280805 | Adjustable 50 mL Conical Tube Rack | 24 x 30 mm | 1 unit |
| 30280798 | Spring Platform | 900 x 540 mm | 1 unit |
| 15361125 | Spring Wire Rack | 240 x 350 mm | Each |
| 30280790 | Spring Platform | 720 x 440 mm | 1 unit |
| 15371125 | Sticky Mat | 140 x 140 mm | Each |
| 30280793 | Spring Platform | 350 x 240 mm | 1 unit |
| 15381125 | Control Box | 158 x 95 mm | Each |
| 30280801 | Spring Platform | 450 x 400 mm | 1 unit |
| 15391125 | General Power Adapter | N/A | Each |
| 30280788 | Spring Platform | 428 x 295 mm | 1 unit |
| 30280789 | Fixed Conical Tube Rack | N/A | 1 unit |
| 30280799 | Adjustable 50 mL Conical Tube Rack | N/A | 1 unit |
| 15301135 | EU Plug | N/A | Each |
| 30280802 | Adjustable 15 mL Conical Tube Rack | N/A | 1 unit |
| 15311135 | UK Plug | N/A | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
- Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
- Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifica
Specifica
| Product Type | 96-well Microplate Holder |
| Dimensions (L x W) | N/A |
| For Use With (Equipment) | CO2 Resistant Shaker |
| No. of Positions | 6 |
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?