Learn More
Marienfeld Superior™ Racks for 12 microscope slides
Description
- Made of plexiglass
- With numbered slots and marking area
- Offers space for up to 12 microscope slides 76x26 mm
Specifications
Specifications
| Slide Capacity | 12 |
| Description | Racks for microscope slides |
| Device Holds | Slides |
| Material | Plexiglass |