Learn More
Hamilton™ Filtered Pre-Sterilized Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
Brand:  Hamilton™ 68007-13
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15375094
  • 1945.00 NOK / Pack of 10
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
  • Pre-sterilized
  • 96 tips to a rack
  • Standard retention
Specifications
Specifications
10 μL
Filtered
Sterile
Polypropylene