Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Available pre-sterilized or non-sterile
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Quantity | 10 racks; 960 total tips |
| Material | Virgin Polypropylene |