Learn More
Spectrum Labs™ Rack Set for CF-2 Fraction Collector
Product Code. 11969578
Click to view available options
For Use With (Application):
Holds 116 x 17 mm or 18 mm diameter tubes
Holds 42 x 28 mm diameter scintilation vials
Unit Size:
Each
Product Code. 11969578
Brand: Spectrum Labs™ 124856
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spectrum Labs™ Rack Set for CF-2 Fraction Collector is a full set of racks for one specifically sized tube designed for use with the versatile CF-2 collector.
  • Designed for use with the versatile CF-2 Fraction Collector
  • Complete set of tube racks designed to accommodate the tubes chosen
Specifications
Type Fraction Collector Tube Rack
For Use With (Application) Holds 116 x 17 mm or 18 mm diameter tubes
For Use With CF-2 Fraction Collector