Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 45 mm Shelf Height
Shop All TENAK Products
Click to view available options
Holds:
Microtiterplates
Unit Size:
Each
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Specifications
| Holds | Microtiterplates |
| Material | Aluminum |
| Positions | 9 |
| For Use With (Equipment) | Chest Frseezers |
| Dimensions (L x W x H) | 135 x 92 x 429 mm |
| Height (Metric) | 45 mm |