Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 65 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
Brand: Tenak™ TE15198
Additional Details : Weight : 1.60000kg
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Microtiterplates (8) | |
Chest Frseezers | |
65 mm |
Aluminum | |
135 x 92 x 543 mm |