Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 45 mm Shelf Height
Product Code. 12468180 Shop All TENAK Products
Click to view available options
Holds:
Microtiterplates
Unit Size:
Each
Product Code. 12468180
Brand: Tenak™ TE15164
Only null left
Add to basket
Add to basket
Eco AluRack for chest freezers is designed to hold microtiterplates.
  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Holds Microtiterplates
Material Aluminum
Positions 8
For Use With (Equipment) Chest Frseezers
Dimensions (L x W x H) 135 x 92 x 383 mm
Height (Metric) 45 mm