Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 45 mm Shelf Height
Product Code. 12458180 Shop All TENAK Products
Click to view available options
Holds:
Microtiterplates
Unit Size:
Each
Product Code. 12458180
Brand: Tenak™ TE15162
Only null left
Add to basket
Add to basket
Eco AluRack for chest freezers is designed to hold microtiterplates.
  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Holds Microtiterplates
Material Aluminum
Positions 7
For Use With (Equipment) Chest Frseezers
Dimensions (L x W x H) 135 x 92 x 336 mm
Height (Metric) 45 mm