Learn More
Tenak™ Stainless Steel Comfort Racks for Chest Freezers
Product Code. 15290794 Shop All TENAK Products
Click to view available options
Holds:
Boxes (10)
Boxes (14)
Boxes (15)
Unit Size:
Each
Product Code. 15290794
Brand: Tenak™ TE24465
Only null left
Add to basket
Add to basket
With spring locks mounted on each shelf to secure cryoboxes. Tenak™ Stainless Steel Comfort Racks for Chest Freezers can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate.
  • Available in a range of heights
  • Can be used with all freezer models
  • Sold without cryoboxes
Specifications
Includes Spring Lock
Product Type Comfort Rack
Holds Boxes (15)
Material Stainless Steel
For Use With (Equipment) Chest freezer
Quantity 1 Pack
Dimensions (L x W x H) 140 x 140 x 540 mm
Width (Metric) 140 mm
Height (Metric) 540 mm
Height (Metric) Shelf 32 mm
Depth (Metric) 140 mm