Description
freezer, side-up, for 5 boxes 32mm high, stainless steel, Tenak
Specifications
Specifications
| Cabinet Material | Stainless Steel |
| For Use With (Equipment) | Chest and upright freezers |
| Array | 1 x 5 |
| Dimensions (DxWxH) Exterior | 695 x 36 x 140 mm |
| Depth (Metric) Exterior | 695 mm |
| Height (Metric) Exterior | 140 mm |
| Width (Metric) Exterior | 36 mm |
| Holds | Cryoboxes (5, 32 mm H) |