Description
freezer, for 20 boxes, 32mm high, stainless steel, with locking rod, Tenak
Specifications
Specifications
| Cabinet Material | Stainless Steel |
| For Use With (Equipment) | All freezer models |
| Includes | Locking rod to secure cryoboxes |
| Array | 1 x 20 |
| Dimensions (DxWxH) Exterior | 140 x 140 x 714 mm |
| Depth (Metric) Exterior | 140 mm |
| Height (Metric) Exterior | 714 mm |
| Width (Metric) Exterior | 140 mm |
| Holds | Cryoboxes (20, 32 mm H) |