missing translation for 'onlineSavingsMsg'
Learn More

Racanisodamine, MedChemExpress

Product Code. p-200095940
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16452396

Brand: Medchem Express HYN2064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Racanisodamine is one of the racemic isomers of anisodamine, resembles anisodamine in pharmacological effect. Racanisodamine is a non-selective muscarinic antagonist, used as a component of eye drops for myopic control.

Racanisodamine is one of the racemic isomers of anisodamine, resembles anisodamine in pharmacological effect. Racanisodamine is a non-selective muscarinic antagonist, used as a component of eye drops for myopic control.

Specifications

Chemical Name or Material Racanisodamine
CAS 17659-49-3
Color White
Purity Grade Notes Research
Molecular Formula C17H23NO4
Quantity 500 mg
Solubility Information DMSO : ≥ 125 mg/mL (409.34 mM)
SMILES O=C(OC1CC(N2C)CC(O)C2C1)C(CO)C3=CC=CC=C3
Molecular Weight (g/mol) 305.37
Formula Weight 305.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.