Learn More
rac N'-Nitrosonornicotine-d4 with (R,S)-N-Nitroso Anabasine-d4 (1:1) (Solution in Acetonitrile 1mg/mL), TRC
Specifications
Specifications
| Chemical Name or Material | (RS)-N-Nitrosoanabasine pyridinyl-d4∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4) |
| CAS | 1020719-68-9∣66148-19-4 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | C10 2H4 H9 N3 O∣C9 2H4 H7 N3 O |
| Quantity | 2 mL |
| Synonym | 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine, 2,3,4,6-tetradeuterio-5-(1-nitrosopyrrolidin-2-yl)pyridine |
| InChI Formula | InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/i3D,4D,6D,8D∣InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2/i1D,3D,5D,7D |
| SMILES | [2H]c1nc([2H])c(C2CCCCN2N=O)c([2H])c1[2H]∣[2H]c1nc([2H])c(C2CCCN2N=O)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine∣2,3,4,6-tetradeuterio-5-(1-nitrosopyrrolidin-2-yl)pyridine |
| Molecular Weight (g/mol) | 195.25∣181.23 |
| Show More |