Learn More
(Rac)-BL-918, MedChemExpress
Product Code. 16495188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495188 10 mg 10mg
16491407 50 mg 50mg
16447917 5 mg 5mg
16435886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16495188 Supplier Medchem Express Supplier No. HY124729A
MedChemExpress (Rac)-BL-918 is the racemate of BL-918. BL-918 is a potent activator of UNC-51-like kinase 1 (ULK1), inducing cytoprotective autophagy for Parkinson’s disease treatment.
(Rac)-BL-918 is the racemate of BL-918. BL-918 is a potent activator of UNC-51-like kinase 1 (ULK1), inducing cytoprotective autophagy for Parkinson’s disease treatment.
Specifications
Chemical Name or Material (Rac)-BL-918
CAS 2435589-07-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H15F8N3OS
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (234.33 mM)
SMILES O=C(NC1=CC=C(F)C=C1F)C(NC(NC2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2)=S)C3=CC=CC=C3
Molecular Weight (g/mol) 533.44
Formula Weight 533.44
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration