Learn More
rac 7-Methoxy Propranolol, TRC
Specifications
Specifications
| Chemical Name or Material | rac 7-Methoxy Propranolol |
| CAS | 76275-53-1 |
| Molecular Formula | C17 H23 N O3 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C17H23NO3/c1-12(2)18-10-14(19)11-21-17-6-4-5-13-7-8-15(20-3)9-16(13)17/h4-9,12,14,18-19H,10-11H2,1-3H3 |
| SMILES | COc1ccc2cccc(OCC(O)CNC(C)C)c2c1 |
| IUPAC Name | 1-(7-methoxynaphthalen-1-yl)oxy-3-(propan-2-ylamino)propan-2-ol |
| Molecular Weight (g/mol) | 289.37 |
| Formula Weight | 289.1678 |