Learn More
rac 7-Methoxy Propranolol, TRC
Product Code. 30471965
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471965 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30471965 Supplier Toronto Research Chemicals Supplier No. TRCM2671001MG
 Restricted Item
Estimated Shipment: 05-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material rac 7-Methoxy Propranolol
CAS 76275-53-1
Molecular Formula C17 H23 N O3
Quantity 1 mg
InChI Formula InChI=1S/C17H23NO3/c1-12(2)18-10-14(19)11-21-17-6-4-5-13-7-8-15(20-3)9-16(13)17/h4-9,12,14,18-19H,10-11H2,1-3H3
SMILES COc1ccc2cccc(OCC(O)CNC(C)C)c2c1
IUPAC Name 1-(7-methoxynaphthalen-1-yl)oxy-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 289.37
Formula Weight 289.1678