missing translation for 'onlineSavingsMsg'
Learn More

Rabusertib, MedChemExpress

Product Code. p-200095802
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16498737 5 mg 5mg
16476138 10 mg 10mg
16452277 50 mg 50mg
16456526 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16498737

Brand: Medchem Express HY14720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM.

Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM.

Specifications

Chemical Name or Material Rabusertib
CAS 911222-45-2
Color White
Purity Grade Notes Research
Molecular Formula C18H22BrN5O3
Quantity 5 mg
Synonym LY2603618 IC-83
Solubility Information DMSO : 31.25 mg/mL (71.63 mM; ultrasonic and warming and heat to 60°C)
SMILES BrC(C=C1NC(NC2=NC=C(N=C2)C)=O)=C(C=C1OC[C@H]3OCCNC3)C
Molecular Weight (g/mol) 436.3
Formula Weight 436.3
Percent Purity 97.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.