missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Nucleoprotein Antibody (12) [HRP] - (CVS-11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33166H
3791.00 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Rabies Virus Nucleoprotein Monoclonal antibody specifically detects Rabies Virus Nucleoprotein in Virus samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| Rabies Virus Nucleoprotein | |
| Monoclonal | |
| HRP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Sandwich ELISA Detection | |
| 12 | |
| PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabies Virus Nucleoprotein (Uniprot#: Q8JXF6; Met1-Ser450). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu