missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabex-5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30019777
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019777 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019777 Supplier Invitrogen™ Supplier No. PA5148258

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rabex-5 Polyclonal Antibody for ICC/IF

RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabex-5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene RABGEF1
Gene Accession No. Q9UJ41
Gene Alias BTB/POZ domain-containing protein KCTD7; RAB guanine nucleotide exchange factor (GEF) 1; RAB guanine nucleotide exchange factor 1; Rab5 exchange factor; rab5 GDP/GTP exchange factor; Rab5 GDP/GTP exchange factor, Rabex5; Rab5 guanine nucleotide exchange factor Rabex-5; Rab5ef; Rabaptin-5-associated exchange factor for Rab5; Rabex5; rabex-5; RABEX5 protein; RABGEF1; RAP1; Ras negative regulator Rabex-5; Ras negative regulator Rabex-5/Rin2; Rin2; zgc:65944
Gene Symbols RABGEF1
Host Species Rabbit
Immunogen E. coli-derived Human RABGEF1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.