Learn More
ACROBiosystems Rabbit CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Flag Tag (MALS verified)
Brand: ACROBiosystems CDD-R52D7-100ug
Additional Details : Weight : 0.00010kg
Description
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Rabbit CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 16.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D is fused wiSpecifications
16.5 kDa/14.5 kDa | |
100 µg | |
Gln 22 - Thr 120 (CD3E) & Thr 22 - Ser 107 (CD3D) | |
CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |
>95% as determined by SDS-PAGE. | |
Rabbit CD3E&CD3D; Heterodimer Protein, His Tag&Flag; Tag (CDD-R52D7) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Thr 120 (CD3E) & Thr 22 - Ser 107 (CD3D) (Accession # Q9TUF9-1 (CD3E) & A0A5F9D3I4-1 (CD3D)). | |
-20°C | |
CD3E & CD3D | |
His Tag & Flag Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
Rabbit |