missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYN1 (phospho S603), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1.
Brand: Abnova PAB9658.100uL
This item is not returnable.
View return policy
Specifications
| SYN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| Syn1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S603 of rat Syn1. | |
| 100 μL | |
| Primary | |
| Bovine, Frog, Human, Mouse, Rat, Zebrafish | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Syn1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24949 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction