Learn More
Rabbit anti-Sheep IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Specifications
Specifications
| Antigen | Rabbit anti-Sheep IgM mu heavy chain |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-sheep IgG recognizes sheep IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide) |
| Host Species | Rabbit |
| Immunogen | Sheep IgM mu heavy chain |
| Purification Method | This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. |
| Show More |
For Research Use Only