Learn More
Rabbit anti-Sheep IgM Fc Secondary Antibody, Novus Biologicals™
Product Code. 18681799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
18681799 10 mg 10mg
1 options
Product Code. 18681799
Brand: Novus Biologicals NBP269753
Rabbit Polyclonal Antibody
IgM Fc Polyclonal antibody specifically detects IgM Fc in Sheep samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM Fc
Applications Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Lyophilized from PBS (pH 7.2)
Host Species Rabbit
Immunogen Purified normal IgM isolated from pooled sheep serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized