Learn More
Shaker, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Shaker.
Brand: Abnova PAB12776.100ug
Additional Details : Weight : 0.02000kg
Specifications
Shaker | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Shaker. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
A synthetic peptide corresponding to N-terminus of Shaker. | |
RUO | |
This antibody recognizes ∼73 KDa of Shaker protein. | |
Store at 4°C. For long term storage store at -20°C to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Drosophila, Human | |
Liquid |
For Research Use Only