missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD-95, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PSD-95.
Brand: Abnova PAB9623.100uL
This item is not returnable.
View return policy
Specifications
| PSD-95 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PSD-95. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus rat PSD-95. | |
| 100 μL | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| Bovine, Human, Mouse, Primate, Rat, Zebrafish | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction