Learn More
Rabbit anti-Mouse IgM Fc Secondary Antibody, HRP, Novus Biologicals™
Product Code. 18648396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
18648396 1 mL 1mL
1 options
Product Code. 18648396
Brand: Novus Biologicals NBP269729
Rabbit Polyclonal Antibody
IgM Fc Polyclonal antibody specifically detects IgM Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Dot Blot, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM Fc
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Dot Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Formulation Lyophilized from PBS (pH 7.2)
Host Species Rabbit
Immunogen Highly purified homogenous IgM isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized