Learn More
Rabbit anti-Mouse IgG2b Secondary Antibody, Novus Biologicals™
Product Code. 18658958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
18658958 10 mg 10mg
1 options
Product Code. 18658958
Brand: Novus Biologicals NBP269599
Rabbit Polyclonal Antibody
IgG2b Polyclonal antibody specifically detects IgG2b in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2b
Applications Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Formulation Lyophilized from PBS (pH 7.2)
Host Species Rabbit
Immunogen Pools of purified homogenous IgG2b isolated from pooled mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized