Learn More
Rabbit anti-Llama IgG (H+L) Secondary Antibody, Novus Biologicals™
Product Code. 18200434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
18200434 1 mg 1mg
1 options
Product Code. 18200434
Brand: Novus Biologicals NBP260696
Rabbit Polyclonal Antibody
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG (H+L)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:30000
Formulation PBS (Phosphate Buffered Saline) pH 7.4 with 0.02% Sodium Azide
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Gene Symbols IGHG1
Host Species Rabbit
Immunogen Purified Llama IgG
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity This rabbit anti-llama secondary antibody recognizes both heavy and light chains
Target Species Llama
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.