Learn More
Invitrogen™ Rabbit anti-Human IgM (Heavy chain) Secondary Antibody, HRP
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP). Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin human serum immunoglobulins, light chains on all human immunoglobulins.
Specifications
Specifications
| Antigen | Human IgM (Heavy chain) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Human Mu immunoglobin. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.