missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/2 (phospho T202/Y204), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERK1/2.
Brand: Abnova PAB16949.100ug
This item is not returnable.
View return policy
Specifications
| ERK1/2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERK1/2. | |
| In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| P27361 | |
| Synthetic phosphopeptide corresponding to residues surrounding T202/Y204 of human ERK1/2. | |
| RUO | |
| This antibody detects endogenous levels of ERK1/2 only when phosphorylated at Thr202/Tyr204. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction