missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DNAH7.
Brand: Abnova PAB22820.100uL
This item is not returnable.
View return policy
Description
DNAH7 is a component of the inner dynein arm of ciliary axonemes (Zhang et al., 2002 [PubMed 11877439]).[supplied by OMIM
Sequence: LKLRCERFVEELESYAKQSEEFYSFGDLQDVQRYLKKAQILNGKLDLAADKIEQFNAEEEAFGWLPSVYPQRKKIQDGLNPYLRLYETAVEFSSNYSpecifications
| DNAH7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DNAH7. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8WXX0 | |
| DNAH7 | |
| Recombinant protein corresponding to amino acids of human DNAH7. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| DNAH7 | |
| DKFZp686C09101/FLJ37196/KIAA0944/MGC39580 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56171 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction