missing translation for 'onlineSavingsMsg'
Learn More

RAB6B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10340-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16896663

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
Specifications

Specifications

RAB6B
Polyclonal
Unconjugated
RAB6B
RAB6B, Ras related protein Rab 6B
Rabbit
Antigen Affinity Chromatography
RUO
270192, 363123, 51560
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61294, Q9NRW1
Rab6b
RAB6B Fusion Protein Ag0322
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers