missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB4B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15603425
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15603425 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15603425 Supplier Invitrogen™ Supplier No. PA549124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with zebrafish, mouse and rat based on sequence homology.

Protein transport. Probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB4B
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB4B
Gene Accession No. P61018
Gene Alias 1500031G17Rik; PP1596; Rab4; RAB4B; RAB4B, member RAS oncogene family; ras-related GTP-binding protein 4b; ras-related protein Rab-4B; small GTP binding protein RAB4B
Gene Symbols RAB4B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 29-58 amino acids from the N-terminal region of human RAB4B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.