missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17827-1-AP-20UL
This item is not returnable.
View return policy
Description
Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.Specifications
| RAB3IL1 | |
| Polyclonal | |
| Unconjugated | |
| RAB3IL1 | |
| GRAB, RAB3IL1, Rabin3 like 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171452, 5866, 74760 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TBN0, Q8VDV3, Q99NH3 | |
| Rab3il1 | |
| RAB3IL1 Fusion Protein Ag12204 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction