missing translation for 'onlineSavingsMsg'
Learn More

RAB3IL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16847824 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847824 150 μL 150µL
16857824 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16847824

Brand: Proteintech 178271AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.

Specifications

Antigen RAB3IL1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB3IL1
Gene Accession No. Q8TBN0, Q8VDV3, Q99NH3
Gene Alias GRAB, RAB3IL1, Rabin3 like 1
Gene Symbols Rab3il1
Host Species Rabbit
Immunogen RAB3IL1 Fusion Protein Ag12204
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171452, 5866, 74760
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.