missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15029-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.Specifications
RAB3A | |
Polyclonal | |
Unconjugated | |
RAB3A | |
RAB3A, Ras related protein Rab 3A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19339, 25531, 5864 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20336, P63011, P63012 | |
RAB3A | |
RAB3A Fusion Protein Ag1903 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |